4'-tert-Butyl-2,2,2-trifluoracetophenon 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(4-(Tert-butyl)phenyl)-2,2,2-trifluoroethanone

PC2501-1G
APOSPC2501-1GEA 198 EUR
APOSPC2501-1G
4'-tert-Butyl-2,2,2-trifluoracetophenon 97%
4'-tert-Butyl-2,2,2-trifluoracetophenon
Formel: C₁₂H₁₃F₃O
Lagertemperatur: Raumtemperatur
MDL: MFCD01319979
CAS-Nummer: 73471-97-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR