(Benzotriazol-1-yloxy)tris(dimethylamino)phosphoniumhexafluorophosphat (BOP)

Lieferant: Apollo Scientific
Gefahr

Synonyme: 1H-Benzotriazol-1-yloxytris(dimethylamino)phosphonium hexafluorophosphate, ((1H-Benzo[d][1,2,3]triazol-1-yl)oxy)tris(dimethylamino)phosphonium hexafluorophosphate(V), BOP reagent, Benzotriazol-1-yloxytris(dimethylamino)-phosphonium hexafluorophosphate

PC2519-25G PC2519-5G PC2519-100G
APOSPC2519-25GEA 21.9 EUR
APOSPC2519-25G APOSPC2519-5G APOSPC2519-100G
(Benzotriazol-1-yloxy)tris(dimethylamino)phosphoniumhexafluorophosphat (BOP)
(Benzotriazol-1-yloxy)tris(dimethylamino)phosphoniumhexafluorophosphat (BOP)

Peptide coupling reagent.

Formel: C₁₂H₂₂F₆N₆OP₂
Schmelzpunkt: 136…140 °C (Zersetzung)
Lagertemperatur: Kühlschrank
MDL: MFCD00011948
CAS-Nummer: 56602-33-6
EINECS: 260-279-1
UN: 1325
ADR: 4.1,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR