(Benzotriazol-1-yloxy)tris(dimethylamino)phosphoniumhexafluorophosphat (BOP)
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
1H-Benzotriazol-1-yloxytris(dimethylamino)phosphonium hexafluorophosphate, ((1H-Benzo[d][1,2,3]triazol-1-yl)oxy)tris(dimethylamino)phosphonium hexafluorophosphate(V), BOP reagent, Benzotriazol-1-yloxytris(dimethylamino)-phosphonium hexafluorophosphate
PC2519-25G
PC2519-5G
PC2519-100G
APOSPC2519-25GEA
21.9
EUR
APOSPC2519-25G
APOSPC2519-5G
APOSPC2519-100G
(Benzotriazol-1-yloxy)tris(dimethylamino)phosphoniumhexafluorophosphat (BOP)
(Benzotriazol-1-yloxy)tris(dimethylamino)phosphoniumhexafluorophosphat (BOP)
Peptide coupling reagent.
Formel:
C₁₂H₂₂F₆N₆OP₂ Schmelzpunkt: 136…140 °C (Zersetzung) Lagertemperatur: Kühlschrank |
MDL:
MFCD00011948 CAS-Nummer: 56602-33-6 EINECS: 260-279-1 UN: 1325 ADR: 4.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...