2-(Phenylthio)-5-(trifluoromethyl)benzoic acid
Lieferant: Apollo Scientific
|
Achtung
|
PC2524-1G
APOSPC2524-1GEA
308
EUR
APOSPC2524-1G
2-(Phenylthio)-5-(trifluoromethyl)benzoic acid
2-(Phenylthio)-5-(trifluoromethyl)benzoic acid
Formel:
C₁₄H₉F₃O₂S Schmelzpunkt: 153…154 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00069137 CAS-Nummer: 52548-96-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...