4,4'-Difluorbenzil 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Bis(4-fluorphenyl)ethandion
PC2534-25G
PC2534-5G
APOSPC2534-25GEA
183
EUR
APOSPC2534-25G
APOSPC2534-5G
4,4'-Difluorbenzil 98%
4,4'-Difluorbenzil
Formel:
C₁₄H₈F₂O₂ Schmelzpunkt: 120…122 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00134541 CAS-Nummer: 579-39-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...