2',6'-Dichloro-4'-(trifluoromethyl)acetophenone 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(2,6-Dichloro-4-(trifluoromethyl)phenyl)ethanone

PC2552E-250MG
APOSPC2552E-250MGEA 26.1 EUR
APOSPC2552E-250MG
2',6'-Dichloro-4'-(trifluoromethyl)acetophenone 97%
2',6'-Dichloro-4'-(trifluoromethyl)acetophenone
Formel: C₉H₅Cl₂F₃O
Molecular Weight: 257.037 g/mol
Siedepunkt: 58 °C (0.6 mm Hg)
Lagertemperatur: Raumtemperatur
MDL: MFCD00221015
CAS-Nummer: 175205-88-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR