2,6-Dichlor-4-(trifluormethyl)benzonitril 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC2554E-1G
APOSPC2554E-1GEA
125
EUR
APOSPC2554E-1G
2,6-Dichlor-4-(trifluormethyl)benzonitril 97%
2,6-Dichlor-4-(trifluormethyl)benzonitril
Formel:
C₈H₂Cl₂F₃N Molecular Weight: 240,100 g/mol Siedepunkt: 66 °C/1mmHg Lagertemperatur: Raumtemperatur |
MDL:
MFCD00220879 CAS-Nummer: 157021-61-9 UN: 3276 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...