2,3-Difluor-4-nitroanisol 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC2555-1G
PC2555-25G
PC2555-5G
APOSPC2555-1GEA
29
EUR
APOSPC2555-1G
APOSPC2555-25G
APOSPC2555-5G
2,3-Difluor-4-nitroanisol 98%
2,3-Difluor-4-nitroanisol
Formel:
C₇H₅F₂NO₃ Molecular Weight: 189.12 g/mol Schmelzpunkt: 96…99 °C Flammpunkt: >110 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD07995055 CAS-Nummer: 66684-59-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...