4,8-Dichlor-2-(trifluormethyl)chinolin 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC2556X-5G
APOSPC2556X-5GEA
102
EUR
APOSPC2556X-5G
4,8-Dichlor-2-(trifluormethyl)chinolin 97%
4,8-Dichloro-2-(trifluoromethyl)quinoline
Formel:
C₁₀H₄Cl₂F₃N Molecular Weight: 266,0467 g/mol Schmelzpunkt: 60…63 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00276590 CAS-Nummer: 18706-35-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...