2-(Trifluoromethoxy)terephthalonitrile 98%

Lieferant: Apollo Scientific

PC2562-1G PC2562-5G PC2562-10G
APOSPC2562-1GEA 0 EUR
APOSPC2562-1G APOSPC2562-5G APOSPC2562-10G
2-(Trifluoromethoxy)terephthalonitrile 98%
2-(Trifluoromethoxy)terephthalonitrile
Formel: C₉H₃F₃N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD00203479
CAS-Nummer: 175278-16-7
UN: 3439
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR