3-Chlor-2,4-difluorbenzaldehyd 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC2571-5G
APOSPC2571-5GEA
85.7
EUR
APOSPC2571-5G
3-Chlor-2,4-difluorbenzaldehyd 97%
3-Chloro-2,4-difluorobenzaldehyde
Formel:
C₇H₃ClF₂O Molecular Weight: 176,55 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04116001 CAS-Nummer: 127675-46-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...