2',3'-Difluoracetophenon 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-(2,3-Difluorphenyl)ethan-1-on
PC2574-100G
PC2574-25G
PC2574-5G
APOSPC2574-100GEA
287
EUR
APOSPC2574-100G
APOSPC2574-25G
APOSPC2574-5G
2',3'-Difluoracetophenon 98%
2',3'-Difluoracetophenon
Formel:
C₈H₆F₂O Siedepunkt: 85 °C/20mmHg Lagertemperatur: Raumtemperatur |
MDL:
MFCD00061274 CAS-Nummer: 18355-80-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...