Lithiumperfluor-1-butansulfonat 95%

Lieferant: Apollo Scientific
Achtung

PC2589-25G
APOSPC2589-25GEA 59.6 EUR
APOSPC2589-25G
Lithiumperfluor-1-butansulfonat 95%
Lithiumperfluor-1-butansulfonat
Formel: C₄F₉LiO₃S
Lagertemperatur: Raumtemperatur
MDL: MFCD03094160
CAS-Nummer: 131651-65-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR