3-Chlor-2,4-difluorbenzotrifluorid 98%

Lieferant: Apollo Scientific
Achtung

PC2597-250MG
APOSPC2597-250MGEA 58.2 EUR
APOSPC2597-250MG
3-Chlor-2,4-difluorbenzotrifluorid 98%
3-Chlor-2,4-difluorbenzotrifluorid
Formel: C₇H₂ClF₅
Molecular Weight: 216,54 g/mol
Siedepunkt: 131…132 °C
Lagertemperatur: Raumtemperatur
CAS-Nummer: 120770-03-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR