4-Methyl-2-[4-(trifluoromethyl)phenyl]-1,3-thiazole-5-carboxylic acid 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC2605-5G
APOSPC2605-5GEA
62.6
EUR
APOSPC2605-5G
4-Methyl-2-[4-(trifluoromethyl)phenyl]-1,3-thiazole-5-carboxylic acid 97%
4-Methyl-2-[4-(trifluoromethyl)phenyl]-1,3-thiazole-5-carboxylic acid
Formel:
C₁₂H₈F₃NO₂S Schmelzpunkt: 237…238 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00068105 CAS-Nummer: 144059-86-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...