3-Brom-5-(trifluormethoxy)anilin 99%

Lieferant: Apollo Scientific
Achtung

PC2614-1G PC2614-5G PC2614-10G
APOSPC2614-1GEA 0 EUR
APOSPC2614-1G APOSPC2614-5G APOSPC2614-10G
3-Brom-5-(trifluormethoxy)anilin 99%
3-Brom-5-(trifluormethoxy)anilin
Formel: C₇H₅BrF₃NO
Lagertemperatur: Raumtemperatur
MDL: MFCD08458076
CAS-Nummer: 914636-35-4
UN: 2810
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR