2,3-Difluorbenzamid 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,3-Difluorbenzoesäureamid

PC2615M-1G
APOSPC2615M-1GEA 26.1 EUR
APOSPC2615M-1G
2,3-Difluorbenzamid 97%
2,3-Difluorbenzamid
Formel: C₇H₅F₂NO
Molecular Weight: 157,12 g/mol
Schmelzpunkt: 114…119 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00061137
CAS-Nummer: 18355-75-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR