5-Bromo-2-fluorophenyl methyl sulphone 99%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Bromo-1-fluoro-2-(methylsulphonyl)benzene
PC2624-1G
PC2624-5G
APOSPC2624-1GEA
116
EUR
APOSPC2624-1G
APOSPC2624-5G
5-Bromo-2-fluorophenyl methyl sulphone 99%
5-Bromo-2-fluorophenyl methyl sulphone
Formel:
C₇H₆BrFO₂S Molecular Weight: 253.089 g/mol Schmelzpunkt: 95…96 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08458078 CAS-Nummer: 914636-38-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...