5-Bromo-2-fluorophenyl methyl sulphone 99%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Bromo-1-fluoro-2-(methylsulphonyl)benzene

PC2624-1G PC2624-5G
APOSPC2624-1GEA 116 EUR
APOSPC2624-1G APOSPC2624-5G
5-Bromo-2-fluorophenyl methyl sulphone 99%
5-Bromo-2-fluorophenyl methyl sulphone
Formel: C₇H₆BrFO₂S
Molecular Weight: 253.089 g/mol
Schmelzpunkt: 95…96 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08458078
CAS-Nummer: 914636-38-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR