4-Fluoro-3-(methylsulphonyl)benzonitrile 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Cyano-2-fluorophenyl methyl sulphone
PC2627-5G
APOSPC2627-5GEA
495
EUR
APOSPC2627-5G
4-Fluoro-3-(methylsulphonyl)benzonitrile 97%
4-Fluoro-3-(methylsulphonyl)benzonitrile
Formel:
C₈H₆FNO₂S Molecular Weight: 199,202 g/mol Schmelzpunkt: 128,9…130.2 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08458081 CAS-Nummer: 914636-47-8 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...