4-(2,2,2-Trifluoroethoxy)phenylhydrazine hydrochloride

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Hydrazino-4-(2,2,2-trifluoroethoxy)benzene hydrochloride, 4-Hydrazino-β,β,β-trifluorophenetole hydrochloride, (4-(2,2,2-Trifluoroethoxy)phenyl)hydrazine hydrochloride

PC2632-1G
APOSPC2632-1GEA 92.8 EUR
APOSPC2632-1G
4-(2,2,2-Trifluoroethoxy)phenylhydrazine hydrochloride
4-(2,2,2-Trifluoroethoxy)phenylhydrazine hydrochloride
Formel: C₈H₉F₃N₂O·ClH
Molecular Weight: 242.626 g/mol
Schmelzpunkt: 185…186 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09763680
CAS-Nummer: 115171-04-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR