trans-2-(Trifluoromethoxy)cinnamic acid

Lieferant: Apollo Scientific
Achtung

PC2641-1G PC2641-250MG PC2641-5G
APOSPC2641-1GEA 0 EUR
APOSPC2641-1G APOSPC2641-250MG APOSPC2641-5G
trans-2-(Trifluoromethoxy)cinnamic acid
trans-2-(Trifluoromethoxy)cinnamic acid
Formel: C₁₀H₇F₃O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD06205273
CAS-Nummer: 399579-92-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR