1-Bromo-1,2-difluoro-1,4-pentadiene

Lieferant: Apollo Scientific

PC2649-1G
APOSPC2649-1GEA 219 EUR
APOSPC2649-1G
1-Bromo-1,2-difluoro-1,4-pentadiene
1-Bromo-1,2-difluoro-1,4-pentadiene
Formel: C₅H₅BrF₂
Molecular Weight: 183 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06246019
CAS-Nummer: 1730-24-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR