Tris(2,2,2-trifluoroethyl) Phosphate
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Tris(2,2,2-trifluoroethyl)phosphate, Phosphoric acid tris(2,2,2-trifluoroethyl) ester
PC2653-25G
APOSPC2653-25GEA
0
EUR
APOSPC2653-25G
Tris(2,2,2-trifluoroethyl) Phosphate
Tris(2,2,2-trifluoroethyl) Phosphate
Formel:
C₆H₆F₉O₄P Molecular Weight: 344.07 g/mol Siedepunkt: 83…86 °C/11mmHg Schmelzpunkt: –20 °C Dichte: 1.5941 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00233117 CAS-Nummer: 358-63-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...