2-Chloro-3-nitro-6-(trifluoromethyl)pyridine

Lieferant: Apollo Scientific

PC2661-1G
APOSPC2661-1GEA 0 EUR
APOSPC2661-1G
2-Chloro-3-nitro-6-(trifluoromethyl)pyridine
2-Chloro-3-nitro-6-(trifluoromethyl)pyridine
Formel: C₆H₂ClF₃N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09864674
CAS-Nummer: 117519-08-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR