2-(Aminomethyl)-5-fluorobenzeneboronic acid, N-BOC protected 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-{[(tert-Butoxycarbonyl)amino]methyl}-5-fluorobenzeneboronic acid

PC2666-1G
APOSPC2666-1GEA 125 EUR
APOSPC2666-1G
2-(Aminomethyl)-5-fluorobenzeneboronic acid, N-BOC protected 98%
2-(Aminomethyl)-5-fluorobenzeneboronic acid, N-BOC protected
Formel: C₁₂H₁₇BFNO₄
Schmelzpunkt: 98…102 °C
Lagertemperatur: Kühlschrank
MDL: MFCD04115639
CAS-Nummer: 850568-43-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR