3-Fluor-(4-methoxycarbonyl)phenylboronsäure 96%

Lieferant: Apollo Scientific
Achtung

Synonyme: (3-Fluoro-4-(methoxycarbonyl)phenyl)boronic acid, 3-Fluoro-4-(methoxycarbonyl)phenylboronic acid, 3-Fluoro-4-methoxycarbonylphenylboronic acid, 3-Fluoro-4-(methoxycarbonyl)benzeneboronic acid

PC2667-25G
APOSPC2667-25GEA 115 EUR
APOSPC2667-25G
3-Fluor-(4-methoxycarbonyl)phenylboronsäure 96%
3-Fluor-(4-methoxycarbonyl)phenylboronsäure
Formel: C₈H₈BFO₄
Molecular Weight: 197.96 g/mol
Schmelzpunkt: 180…184 °C
Lagertemperatur: Kühlschrank
MDL: MFCD04115665
CAS-Nummer: 505083-04-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR