2-Aminomethyl-5-fluorobenzeneboronic acid hydrochloride

Lieferant: Apollo Scientific

PC2668-1G
APOSPC2668-1GEA 285 EUR
APOSPC2668-1G
2-Aminomethyl-5-fluorobenzeneboronic acid hydrochloride
2-Aminomethyl-5-fluorobenzeneboronic acid hydrochloride
Formel: C₇H₉BFNO₂·HCl
Molecular Weight: 205,42 g/mol
Schmelzpunkt: 152…158 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04115666
CAS-Nummer: 850568-03-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR