2-Amino-5-fluor-3-iodpyridin

Lieferant: Apollo Scientific
Achtung

PC2681-5G
APOSPC2681-5GEA 186 EUR
APOSPC2681-5G
2-Amino-5-fluor-3-iodpyridin
2-Amino-5-fluoro-3-iodopyridine
Formel: C₅H₄FIN₂
Molecular Weight: 238.0015 g/mol
Schmelzpunkt: 70…78 °C
Lagertemperatur: Kühlschrank
MDL: MFCD08688587
CAS-Nummer: 823218-51-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR