4,4'-Difluorbenzophenon 98+%
Lieferant: Apollo Scientific
|
Achtung
|
PC2696-1KG
PC2696-250G
APOSPC2696-1KGEA
109
EUR
APOSPC2696-1KG
APOSPC2696-250G
4,4'-Difluorbenzophenon 98+%
4,4'-Difluorbenzophenon
Intermediate for the manufacture of polyetheretherketone(PEEK) high performance polymer.
Formel:
C₁₃H₈F₂O Siedepunkt: 170…172 °C/10mmHg Schmelzpunkt: 102…110 °C Dichte: 1,39 g/cm³ (25 °C) Flammpunkt: 152 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000353 CAS-Nummer: 345-92-6 EINECS: 206-466-3 UN: 3077 ADR: 9+EHS,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...