4-Chloro-3-(trifluoromethyl)phenylhydrazine hydrochloride

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Chloro-5-hydrazinobenzotrifluoride hydrochloride, 1-Chloro-4-hydrazino-2-(trifluoromethyl)benzene hydrochloride, (4-Chloro-3-(trifluoromethyl)phenyl)hydrazine hydrochloride

PC2711-1G
APOSPC2711-1GEA 52.2 EUR
APOSPC2711-1G
4-Chloro-3-(trifluoromethyl)phenylhydrazine hydrochloride
4-Chloro-3-(trifluoromethyl)phenylhydrazine hydrochloride
Formel: C₇H₆ClF₃N₂·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD00832922
CAS-Nummer: 40566-70-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR