(2-Chloro-5-fluorophenyl)hydrazine hydrochloride

Lieferant: Apollo Scientific
Achtung

PC2716-1G
APOSPC2716-1GEA 21.9 EUR
APOSPC2716-1G
(2-Chloro-5-fluorophenyl)hydrazine hydrochloride
(2-Chloro-5-fluorophenyl)hydrazine hydrochloride
Formel: C₆H₇Cl₂FN₂
Molecular Weight: 197,0400 g/mol
Flammpunkt: >110 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03094167
CAS-Nummer: 502496-25-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR