α-Brom-2,6-difluortoluol 98%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 2,6-Difluorbenzylbromid

PC2743-25G PC2743-100G
APOSPC2743-25GEA 42.2 EUR
APOSPC2743-25G APOSPC2743-100G
α-Brom-2,6-difluortoluol 98%
α-Brom-2,6-difluortoluol

Can protect alcohols as 2,6-difluorobenzyl ethers see : Helv. Chim. Acta, 79, 1159 (1996), these ethersunlike benzyl ethers are stable during cationic reactions.

Formel: C₇H₅BrF₂
Schmelzpunkt: 51…55 °C
Flammpunkt: 113 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00000329
CAS-Nummer: 85118-00-9
EINECS: 285-652-6
UN: 3261
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR