2,3,4-Trifluorophenylhydrazine hydrochloride

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Hydrazino-2,3,4-trifluorobenzene hydrochloride

PC2751-5G
APOSPC2751-5GEA 105 EUR
APOSPC2751-5G
2,3,4-Trifluorophenylhydrazine hydrochloride
2,3,4-Trifluorophenylhydrazine hydrochloride
Formel: C₆H₅F₃N₂·ClH
Molecular Weight: 198.573 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03094176
CAS-Nummer: 502496-30-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR