2-Amino-4-bromo-6-fluorophenol 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Bromo-3-fluoro-2-hydroxyaniline
PC2775-1G
PC2775-5G
APOSPC2775-1GEA
116
EUR
APOSPC2775-1G
APOSPC2775-5G
2-Amino-4-bromo-6-fluorophenol 97%
2-Amino-4-bromo-6-fluorophenol
Formel:
C₆H₅BrFNO Molecular Weight: 206.012 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03094184 CAS-Nummer: 182499-89-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...