2-Amino-4-bromo-6-fluorophenol 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Bromo-3-fluoro-2-hydroxyaniline

PC2775-1G PC2775-5G
APOSPC2775-1GEA 116 EUR
APOSPC2775-1G APOSPC2775-5G
2-Amino-4-bromo-6-fluorophenol 97%
2-Amino-4-bromo-6-fluorophenol
Formel: C₆H₅BrFNO
Molecular Weight: 206.012 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03094184
CAS-Nummer: 182499-89-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR