4,5-Difluor-2-nitrophenol 97%

Lieferant: Apollo Scientific
Achtung

PC2779-1G PC2779-25G PC2779-5G
APOSPC2779-1GEA 21.9 EUR
APOSPC2779-1G APOSPC2779-25G APOSPC2779-5G
4,5-Difluor-2-nitrophenol 97%
4,5-Difluor-2-nitrophenol
Formel: C₆H₃F₂NO₃
Molecular Weight: 175.0897 g/mol
Schmelzpunkt: 54…57 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03094187
CAS-Nummer: 55346-97-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR