N-[1-(Hydrazinocarbonyl)-3-(methylthio)prop-1-yl]-4-(trifluoromethyl)benzamide 97%

Lieferant: Apollo Scientific

Synonyme: N-[1-Hydrazino-4-(methylsulphanyl)-1-oxobut-2-yl]-4-(trifluoromethyl)benzamide, 4-{[1-Hydrazino-4-(methylsulphanyl)-1-oxobut-2-yl]carbamoyl}benzotrifluoride

PC2781-1G
APOSPC2781-1GEA 201 EUR
APOSPC2781-1G
N-[1-(Hydrazinocarbonyl)-3-(methylthio)prop-1-yl]-4-(trifluoromethyl)benzamide 97%
N-[1-(Hydrazinocarbonyl)-3-(methylthio)prop-1-yl]-4-(trifluoromethyl)benzamide
Formel: C₁₃H₁₆F₃N₃O₂S
Molecular Weight: 335,345 g/mol
Schmelzpunkt: 181…184 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00174305
CAS-Nummer: 209056-84-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR