Fmoc-Lys(Tfa)-OH

Lieferant: Apollo Scientific

PC27880-25G
APOSPC27880-25GEA 97.3 EUR
APOSPC27880-25G
Fmoc-Lys(Tfa)-OH
Fmoc-Lys(Tfa)-OH
Formel: C₂₃H₂₃F₃N₂O₅
Lagertemperatur: Raumtemperatur
MDL: MFCD00153360
CAS-Nummer: 76265-69-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR