(S)-2-(9H-Fluoren-9-ylmethoxycarbonylamino)-4,4-difluoro-butyric acid

Lieferant: Apollo Scientific

Synonyme: (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4,4-difluorobutanoic acid

PC27883-250MG
APOSPC27883-250MGEA 136 EUR
APOSPC27883-250MG
(S)-2-(9H-Fluoren-9-ylmethoxycarbonylamino)-4,4-difluoro-butyric acid
(S)-2-(9H-Fluoren-9-ylmethoxycarbonylamino)-4,4-difluoro-butyric acid
Formel: C₁₉H₁₇F₂NO₄
Molecular Weight: 361,339 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06796145
CAS-Nummer: 467442-21-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR