N-Fmoc-6-Fluoro-L-tryptophan

Lieferant: Apollo Scientific

PC27891-100MG PC27891-250MG PC27891-50MG
APOSPC27891-100MGEA 30.7 EUR
APOSPC27891-100MG APOSPC27891-250MG APOSPC27891-50MG
N-Fmoc-6-Fluoro-L-tryptophan
N-Fmoc-6-Fluoro-L-tryptophan
Formel: C₂₆H₂₁FN₂O₄
Lagertemperatur: Raumtemperatur
MDL: MFCD29917530
CAS-Nummer: 908847-01-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR