5-Bromo-2-iodobenzyl fluoride 95%

Lieferant: Apollo Scientific
Gefahr

PC27894-250MG
APOSPC27894-250MGEA 79.9 EUR
APOSPC27894-250MG
5-Bromo-2-iodobenzyl fluoride 95%
5-Bromo-2-iodobenzyl fluoride
Formel: C₇H₅BrFI
Molecular Weight: 314.921 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31664892
CAS-Nummer: 2244088-43-3
UN: 3265
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR