3-Bromo-4-(trifluoromethoxy)benzyl fluoride 95%

Lieferant: Apollo Scientific
Gefahr

PC27902-1G PC27902-250MG
APOSPC27902-1GEA 609 EUR
APOSPC27902-1G APOSPC27902-250MG
3-Bromo-4-(trifluoromethoxy)benzyl fluoride 95%
3-Bromo-4-(trifluoromethoxy)benzyl fluoride
Formel: C₈H₅BrF₄O
Molecular Weight: 273.022 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31664895
CAS-Nummer: 2169407-53-6
UN: 3265
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR