2-Bromo-3-chloro-6-fluorobenzyl fluoride 95%
Lieferant: Apollo Scientific
|
Gefahr
|
PC27913-250MG
APOSPC27913-250MGEA
79.9
EUR
APOSPC27913-250MG
2-Bromo-3-chloro-6-fluorobenzyl fluoride 95%
2-Bromo-3-chloro-6-fluorobenzyl fluoride
Formel:
C₇H₄BrClF₂ Molecular Weight: 241.46 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD31664901 CAS-Nummer: 2244087-33-8 UN: 3261 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...