3-Bromo-4,5-difluorobenzyl fluoride 95%

Lieferant: Apollo Scientific
Gefahr

PC27915-250MG
APOSPC27915-250MGEA 79.9 EUR
APOSPC27915-250MG
3-Bromo-4,5-difluorobenzyl fluoride 95%
3-Bromo-4,5-difluorobenzyl fluoride
Formel: C₇H₄BrF₃
Lagertemperatur: Raumtemperatur
MDL: MFCD31664902
CAS-Nummer: 1785082-71-4
UN: 3265
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR