5-Bromo-2,3-difluorobenzyl fluoride 95%

Lieferant: Apollo Scientific
Gefahr

PC27917-250MG
APOSPC27917-250MGEA 79.9 EUR
APOSPC27917-250MG
5-Bromo-2,3-difluorobenzyl fluoride 95%
5-Bromo-2,3-difluorobenzyl fluoride
Formel: C₇H₄BrF₃
Molecular Weight: 225.006 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31664903
CAS-Nummer: 2244085-63-8
UN: 3265
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR