3-Bromo-2,4,5-trifluorobenzyl fluoride 95%

Lieferant: Apollo Scientific
Gefahr

PC27919-250MG
APOSPC27919-250MGEA 79.9 EUR
APOSPC27919-250MG
3-Bromo-2,4,5-trifluorobenzyl fluoride 95%
3-Bromo-2,4,5-trifluorobenzyl fluoride
Formel: C₇H₃BrF₄
Lagertemperatur: Raumtemperatur
MDL: MFCD31664904
CAS-Nummer: 2244088-42-2
UN: 3265
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR