1-Phenyl-3-(trifluoromethyl)pyrazole 95+%

Lieferant: Apollo Scientific

Synonyme: 1-Phenyl-3-(trifluoromethyl)-1H-pyrazole

PC2792-250MG
APOSPC2792-250MGEA 105 EUR
APOSPC2792-250MG
1-Phenyl-3-(trifluoromethyl)pyrazole 95+%
1-Phenyl-3-(trifluoromethyl)pyrazole
Formel: C₁₀H₇F₃N₂
Molecular Weight: 212,171 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03094197
CAS-Nummer: 99498-65-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR