2-Bromo-5-fluoro-3-nitrobenzyl fluoride 95%

Lieferant: Apollo Scientific
Gefahr

PC27920-1G PC27920-250MG
APOSPC27920-1GEA 609 EUR
APOSPC27920-1G APOSPC27920-250MG
2-Bromo-5-fluoro-3-nitrobenzyl fluoride 95%
2-Bromo-5-fluoro-3-nitrobenzyl fluoride
Formel: C₇H₄BrF₂NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD28738751
CAS-Nummer: 1807004-68-7
UN: 3261
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR