2-Bromo-5-fluoro-3-nitrobenzyl fluoride 95%
Lieferant: Apollo Scientific
|
Gefahr
|
PC27920-1G
PC27920-250MG
APOSPC27920-1GEA
609
EUR
APOSPC27920-1G
APOSPC27920-250MG
2-Bromo-5-fluoro-3-nitrobenzyl fluoride 95%
2-Bromo-5-fluoro-3-nitrobenzyl fluoride
Formel:
C₇H₄BrF₂NO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD28738751 CAS-Nummer: 1807004-68-7 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...